Anti-ITSN2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ITSN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-949
- 77838-948
- Antigen symbol:
- ITSN2
- ITSN2
- Antigen name:
- Intersectin-2 protein (940-1209AA)
- Intersectin-2 protein (940-1209AA)
- Antigen synonyms:
- ITSN2| KIAA1256| SH3D1B| SWAP| Intersectin-2| SH3 domain-containing protein 1B| SH3P18| SH3P18-like WASP-associated protein
- ITSN2| KIAA1256| SH3D1B| SWAP| Intersectin-2| SH3 domain-containing protein 1B| SH3P18| SH3P18-like WASP-associated protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NZM3
- Q9NZM3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together