Anti-SEC24C Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SEC24C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77798-989
- 77798-990
- Antigen symbol:
- SEC24C
- SEC24C
- Antigen name:
- Protein transport protein Sec24C protein (200-480AA)
- Protein transport protein Sec24C protein (200-480AA)
- Antigen synonyms:
- Protein transport protein Sec24C|SC 24C|SC24 C|SC24C|SC24C_HUMAN|Sec 24C|Sec24 C|SEC24 related gene family, member C (S. cerevisiae)|SEC24 related protein C|SEC24-related protein C|SEC24C
- Protein transport protein Sec24C|SC 24C|SC24 C|SC24C|SC24C_HUMAN|Sec 24C|Sec24 C|SEC24 related gene family, member C (S. cerevisiae)|SEC24 related protein C|SEC24-related protein C|SEC24C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P53992
- P53992
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together