Anti-CYFIP1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CYFIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-203
- 77830-202
- Antigen symbol:
- CYFIP1
- CYFIP1
- Antigen name:
- Cytoplasmic FMR1-interacting protein 1 protein (320-449AA)
- Cytoplasmic FMR1-interacting protein 1 protein (320-449AA)
- Antigen synonyms:
- CY| CYFIP 1| CYFIP1| CYFP1_HUMAN| Cytoplasmic FMR1 interacting protein 1| Cytoplasmic FMR1-interacting protein 1| FLJ45151| KIAA0068| p140sra 1| p140sra-1| P140SRA1| Selective hybridizing clone| SHYC| Specifically Rac1 associated protein 1| Specifically Rac1-associated protein 1| SRA 1| Sra-1| SRA1
- CY| CYFIP 1| CYFIP1| CYFP1_HUMAN| Cytoplasmic FMR1 interacting protein 1| Cytoplasmic FMR1-interacting protein 1| FLJ45151| KIAA0068| p140sra 1| p140sra-1| P140SRA1| Selective hybridizing clone| SHYC| Specifically Rac1 associated protein 1| Specifically Rac1-associated protein 1| SRA 1| Sra-1| SRA1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L576
- Q7L576
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together