Anti-UGT1A9 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UGT1A9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-971
- 77803-971
- Antigen symbol:
- UGT1A9
- UGT1A9
- Antigen name:
- UDP-glucuronosyltransferase 1-9 protein (277-473AA)
- UDP-glucuronosyltransferase 1-9 protein (277-473AA)
- Antigen synonyms:
- GNT1|HLUGP4|lugP4|UD19_HUMAN|UDP glucuronosyltransferase 1A9|UDP glucuronosyltransferase family 1 member A9|UDP-glucuronosyltransferase 1 family|polypeptide A9|UDP-glucuronosyltransferase 1-9|UDP-glucuronosyltransferase 1-I|UDP-glucuronosyltransferase 1A9|UDPGT 1-9|UDPGT|UGT-1I|UGT1*9|UGT1-09|UGT1.9|Ugt1a9|UGT1A9S|UGT1AI|UGT1I
- GNT1|HLUGP4|lugP4|UD19_HUMAN|UDP glucuronosyltransferase 1A9|UDP glucuronosyltransferase family 1 member A9|UDP-glucuronosyltransferase 1 family|polypeptide A9|UDP-glucuronosyltransferase 1-9|UDP-glucuronosyltransferase 1-I|UDP-glucuronosyltransferase 1A9|UDPGT 1-9|UDPGT|UGT-1I|UGT1*9|UGT1-09|UGT1.9|Ugt1a9|UGT1A9S|UGT1AI|UGT1I
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O60656
- O60656
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together