Order Entry
Canada
ContactUsLinkComponent
Anti-MAP1LC3C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-MAP1LC3C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MAP1LC3C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77842-964
  • 77842-963
  • Antigen symbol:
  • MAP1LC3C
  • MAP1LC3C
  • Antigen name:
  • Microtubule-associated proteins 1A/1B light chain 3C protein (1-30AA)
  • Microtubule-associated proteins 1A/1B light chain 3C protein (1-30AA)
  • Antigen synonyms:
  • ATG8J|Autophagy-related protein LC3 C|Autophagy-related ubiquitin-like modifier LC3 C|LC3 like protein 2|MAP1 light chain 3 like protein 2|MAP1 light chain 3 like protein 3|MAP1 light chain 3-like protein 3|MAP1A/MAP1B LC3 C|MAP1A/MAP1B light chain 3 C|MAP1LC3C|Microtubule-associated protein 1 light chain 3 gamma|Microtubule-associated proteins 1A/1B light chain 3C|MLP3C_HUMAN
  • ATG8J|Autophagy-related protein LC3 C|Autophagy-related ubiquitin-like modifier LC3 C|LC3 like protein 2|MAP1 light chain 3 like protein 2|MAP1 light chain 3 like protein 3|MAP1 light chain 3-like protein 3|MAP1A/MAP1B LC3 C|MAP1A/MAP1B light chain 3 C|MAP1LC3C|Microtubule-associated protein 1 light chain 3 gamma|Microtubule-associated proteins 1A/1B light chain 3C|MLP3C_HUMAN
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9BXW4
  • Q9BXW4
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together