Anti-KDM4C Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KDM4C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-352
- 77838-351
- Antigen symbol:
- KDM4C
- KDM4C
- Antigen name:
- Lysine-specific demethylase 4C protein (450-600AA)
- Lysine-specific demethylase 4C protein (450-600AA)
- Antigen synonyms:
- bA146B14.1|GASC 1 protein|GASC-1 protein|GASC1|Gene amplified in squamous cell carcinoma 1 protein|JHDM3C|JmjC domain containing histone demethylation protein 3C|JmjC domain-containing histone demethylation protein 3C|JMJD2C|Jumonji domain containing 2C|Jumonji domain containing protein 2C|Jumonji domain-containing protein 2C|Kdm4c|KDM4C_HUMAN|Lysine (K) specific demethylase 4C|Lysine demethylase 4C|Lysine-specific demethylase 4C|TDRD14C|Tudor domain containing 14C
- bA146B14.1|GASC 1 protein|GASC-1 protein|GASC1|Gene amplified in squamous cell carcinoma 1 protein|JHDM3C|JmjC domain containing histone demethylation protein 3C|JmjC domain-containing histone demethylation protein 3C|JMJD2C|Jumonji domain containing 2C|Jumonji domain containing protein 2C|Jumonji domain-containing protein 2C|Kdm4c|KDM4C_HUMAN|Lysine (K) specific demethylase 4C|Lysine demethylase 4C|Lysine-specific demethylase 4C|TDRD14C|Tudor domain containing 14C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H3R0
- Q9H3R0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together