Anti-sdeA Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: sdeA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Legionella pneumophila subsp
Specifications
- Catalog No:
- 77813-737
- 77813-738
- Antigen symbol:
- sdeA
- sdeA
- Antigen name:
- Recombinant Legionella pneumophila subsp Ubiquitinating/deubiquitinating enzyme SdeA protein (19-268AA)
- Recombinant Legionella pneumophila subsp Ubiquitinating/deubiquitinating enzyme SdeA protein (19-268AA)
- Antigen synonyms:
- sdeA|lpg2157|Ubiquitinating/deubiquitinating enzyme SdeA|Effector protein SdeA) [Includes: Deubiquitinase|DUB|EC 3.4.22.-|Deneddylase|Deubiquitinating enzyme)|Ubiquitin transferase|EC 2.3.2.-)|Mono-ADP-ribosyltransferase|mART|EC 2.4.2.31)]
- sdeA|lpg2157|Ubiquitinating/deubiquitinating enzyme SdeA|Effector protein SdeA) [Includes: Deubiquitinase|DUB|EC 3.4.22.-|Deneddylase|Deubiquitinating enzyme)|Ubiquitin transferase|EC 2.3.2.-)|Mono-ADP-ribosyltransferase|mART|EC 2.4.2.31)]
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Legionella pneumophila subsp
- Legionella pneumophila subsp
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5ZTK4
- Q5ZTK4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together