Anti-SLCO1A2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: SLCO1A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77800-178
- 77800-179
- Antigen symbol:
- SLCO1A2
- SLCO1A2
- Antigen name:
- Solute carrier organic anion transporter family member 1A2 protein (406-513AA)
- Solute carrier organic anion transporter family member 1A2 protein (406-513AA)
- Antigen synonyms:
- OATP|OATP-1|OATP-A|OATP1A2|Organic anion-transporting polypeptide 1|SLC21A3|SLCO1A2|SO1A2_HUMAN|Sodium-independent organic anion transporter|Solute carrier family 21 member 3|Solute carrier organic anion transporter family member 1A2
- OATP|OATP-1|OATP-A|OATP1A2|Organic anion-transporting polypeptide 1|SLC21A3|SLCO1A2|SO1A2_HUMAN|Sodium-independent organic anion transporter|Solute carrier family 21 member 3|Solute carrier organic anion transporter family member 1A2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P46721
- P46721
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together