Anti-DCTN6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:2000, WB:1:200 to 1:1000, IHC:1:100 to 1:300
Type:
Antigen: DCTN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77820-612
- 77820-611
- Antigen symbol:
- DCTN6
- DCTN6
- Antigen name:
- Fusion protein of DCTN6
- Fusion protein of DCTN6
- Antigen synonyms:
- DCTN6|DCTN6_HUMAN|Dynactin 6|Dynactin subunit 6|Dynactin subunit p27|Novel RGD containing protein|p27|Protein WS-3|Protein WS3|WS 3|WS3
- DCTN6|DCTN6_HUMAN|Dynactin 6|Dynactin subunit 6|Dynactin subunit p27|Novel RGD containing protein|p27|Protein WS-3|Protein WS3|WS 3|WS3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00399
- O00399
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together