Anti-RBMY1A1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77815-116
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:RNA-binding motif protein, Y chromosome, family 1 member A1 protein (1-459AA)
- Antigen symbol:RBMY1A1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P0DJD3
- Antigen synonyms:family 1 member A1|hRBMY|MGC181956|RBM|RBM1|RBM2|RBMY|Rbmy1a1|RBMY1C|RBY1A_HUMAN|RNA binding motif protein 1|RNA binding motif protein 2|RNA binding motif protein Y chromosome family 1 member A1|RNA binding motif protein Y chromosome family 1 member A1/C|RNA binding motif protein Y chromosome family 1 member|RNA binding motif protein Y linked family 1 member A1|RNA-binding motif protein 1|RNA-binding motif protein 2|RNA-binding motif protein|Y chromosome|Y chromosome RNA recognition motif 1|YRRM1|YRRM2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: RBMY1A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat