Anti-CD109 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: CD109
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77829-517
- 77829-516
- Antigen symbol:
- CD109
- CD109
- Antigen name:
- CD109 antigen protein (967-1122AA)
- CD109 antigen protein (967-1122AA)
- Antigen synonyms:
- 150 KDa TGF beta 1 binding protein|150 KDa TGF-beta-1-binding protein|7D1|8A3|Activated T cell marker CD109|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7|CD 109|CD109|CD109 antigen|CD109_HUMAN|CPAMD7|DKFZp762L1111|E123|FLJ38569|FLJ41966|Gov system alloantigens on platelets|p180|Platelet specific Gov antigen|Platelet-specific Gov antigen|r150|RP11 525G3.1
- 150 KDa TGF beta 1 binding protein|150 KDa TGF-beta-1-binding protein|7D1|8A3|Activated T cell marker CD109|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7|CD 109|CD109|CD109 antigen|CD109_HUMAN|CPAMD7|DKFZp762L1111|E123|FLJ38569|FLJ41966|Gov system alloantigens on platelets|p180|Platelet specific Gov antigen|Platelet-specific Gov antigen|r150|RP11 525G3.1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6YHK3
- Q6YHK3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together