Anti-ACSM3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ACSM3
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77827-768
- 77827-769
- Antigen symbol:
- ACSM3
- ACSM3
- Antigen name:
- Acyl-coenzyme A synthetase ACSM3, mitochondrial protein (387-586AA)
- Acyl-coenzyme A synthetase ACSM3, mitochondrial protein (387-586AA)
- Antigen synonyms:
- ACSM3| ACSM3_HUMAN| Acyl CoA synthetase medium chain family member 3| Acyl-CoA synthetase medium-chain family member 3| Acyl-coenzyme A synthetase ACSM3| mitochondrial| Butyrate CoA ligase 3| Butyrate--CoA ligase 3| Butyryl coenzyme A synthetase 3| Butyryl-coenzyme A synthetase 3| Middle chain acyl CoA synthetase 3| Middle-chain acyl-CoA synthetase 3| Protein SA homolog| SA (rat hypertension associated) homolog| SA| SA hypertension associated homolog| rat| homolog of| SAH
- ACSM3| ACSM3_HUMAN| Acyl CoA synthetase medium chain family member 3| Acyl-CoA synthetase medium-chain family member 3| Acyl-coenzyme A synthetase ACSM3| mitochondrial| Butyrate CoA ligase 3| Butyrate--CoA ligase 3| Butyryl coenzyme A synthetase 3| Butyryl-coenzyme A synthetase 3| Middle chain acyl CoA synthetase 3| Middle-chain acyl-CoA synthetase 3| Protein SA homolog| SA (rat hypertension associated) homolog| SA| SA hypertension associated homolog| rat| homolog of| SAH
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q53FZ2
- Q53FZ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together