Anti-ZRANB3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ZRANB3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77827-555
- 77827-556
- Antigen symbol:
- ZRANB3
- ZRANB3
- Antigen name:
- DNA annealing helicase and endonuclease ZRANB3 protein (370-624AA)
- DNA annealing helicase and endonuclease ZRANB3 protein (370-624AA)
- Antigen synonyms:
- 4933425L19Rik|AH2|Annealing helicase 2|DNA annealing helicase and endonuclease ZRANB3|MGC105033|MGC75012|OTTHUMP00000197449|Zinc finger Ran-binding domain-containing protein 3|Zinc finger|RAN- binding domain containing 3|ZRAB3_HUMAN|Zranb3
- 4933425L19Rik|AH2|Annealing helicase 2|DNA annealing helicase and endonuclease ZRANB3|MGC105033|MGC75012|OTTHUMP00000197449|Zinc finger Ran-binding domain-containing protein 3|Zinc finger|RAN- binding domain containing 3|ZRAB3_HUMAN|Zranb3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5FWF4
- Q5FWF4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together