Anti-TMEM150A Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: TMEM150A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77832-663
- 77832-662
- Antigen symbol:
- TMEM150A
- TMEM150A
- Antigen name:
- Transmembrane protein 150A protein (24-75AA)
- Transmembrane protein 150A protein (24-75AA)
- Antigen synonyms:
- TMEM150A|TMEM150|Transmembrane protein 150A|Transmembrane protein 150
- TMEM150A|TMEM150|Transmembrane protein 150A|Transmembrane protein 150
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86TG1
- Q86TG1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together