Anti-CACNA2D1 Rabbit Polyclonal Antibody
Catalog # 77809-081
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Synthetic peptide corresponding to a region derived from internal residues of calcium channel, voltage-dependent, L type, alpha 1D subunit
- Antigen symbol:CACNA2D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:P54289
- Antigen synonyms:CA2D1_HUMAN|CACN A2|CACNA2|Cacna2d1|CACNL2A|Calcium channel L type alpha 2 polypeptide|Calcium channel voltage dependent alpha 2/delta subunit 1|CCHL2A|Dihydropyridine receptor alpha 2 subunit|Dihydropyridine sensitive L type calcium channel alpha 2/delta subunit|Dihydropyridine sensitive L type calcium channel subunits alpha 2/delta|L type calcium channel subunit alpha 2|MHS 3|MHS3|Voltage dependent calcium channel subunit alpha 2/delta 1|Voltage gated calcium channel subunit alpha 2/delta 1|Voltage-dependent calcium channel subunit delta-1|Voltage-gated calcium channel subunit alpha-2/delta-1
- Storage buffer:PBS, 0.05% Sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended Dilution: IHC: 1:50 to 1:100
Type:
Antigen: CACNA2D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat