Anti-RNF213 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: RNF213
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77828-061
- 77828-062
- Antigen symbol:
- RNF213
- RNF213
- Antigen name:
- E3 ubiquitin-protein ligase RNF213 protein (202-350AA)
- E3 ubiquitin-protein ligase RNF213 protein (202-350AA)
- Antigen synonyms:
- ALK lymphoma oligomerization partner on chromosome 17| C17orf27| Chromosome 17 open reading frame 27| E3 ubiquitin-protein ligase RNF213| FLJ13051| KIAA1554| MGC46622| MGC9929| MYMY2| Mysterin| MYSTR| NET57| RING finger protein 213| RN213_HUMAN| RNF 213| RNF213
- ALK lymphoma oligomerization partner on chromosome 17| C17orf27| Chromosome 17 open reading frame 27| E3 ubiquitin-protein ligase RNF213| FLJ13051| KIAA1554| MGC46622| MGC9929| MYMY2| Mysterin| MYSTR| NET57| RING finger protein 213| RN213_HUMAN| RNF 213| RNF213
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q63HN8
- Q63HN8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together