Anti-SHD Rabbit Polyclonal Antibody
Catalog # 77807-192
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the N-terminal region of Shd.
- Antigen symbol:SHD
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96IW2
- Antigen synonyms:SHD| SH2 domain-containing adapter protein D
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: SHD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human