Anti-SLC32A1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: SLC32A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77819-174
- 77819-173
- Antigen symbol:
- SLC32A1
- SLC32A1
- Antigen name:
- Synthetic peptide of SLC32A1
- Synthetic peptide of SLC32A1
- Antigen synonyms:
- bA122O1.1|GABA and glycine transporter|hVIAAT|SLC32A 1|Slc32a1|solute carrier family 32 (GABA vesicular transporter) member 1|Solute carrier family 32 member 1|Vesicular GABA Amino Acid Transporter|Vesicular GABA transporter|Vesicular inhibitory amino acid transporter|VGAT|VIAAT|VIAAT_HUMAN
- bA122O1.1|GABA and glycine transporter|hVIAAT|SLC32A 1|Slc32a1|solute carrier family 32 (GABA vesicular transporter) member 1|Solute carrier family 32 member 1|Vesicular GABA Amino Acid Transporter|Vesicular GABA transporter|Vesicular inhibitory amino acid transporter|VGAT|VIAAT|VIAAT_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H598
- Q9H598
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together