Anti-ID3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: ID3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77797-092
- 77797-093
- Antigen symbol:
- ID3
- ID3
- Antigen name:
- DNA-binding protein inhibitor ID-3 protein (27-99AA)
- DNA-binding protein inhibitor ID-3 protein (27-99AA)
- Antigen synonyms:
- 1R21|bHLHb25|Class B basic helix-loop-helix protein 25|DNA binding protein inhibitor ID 3|DNA-binding protein inhibitor ID-3|HEIR 1|HEIR1|Helix loop helix protein HEIR 1|Helix loop helix protein HEIR1|Helix-loop-helix protein HEIR-1|HLH642|ID 3|ID like protein inhibitor HLH 1R21|ID like protein inhibitor HLH 462|ID-like protein inhibitor HLH 1R21|ID3|ID3_HUMAN|Inhibitor of DNA binding 3|Inhibitor of DNA binding 3 dominant negative helix loop helix protein
- 1R21|bHLHb25|Class B basic helix-loop-helix protein 25|DNA binding protein inhibitor ID 3|DNA-binding protein inhibitor ID-3|HEIR 1|HEIR1|Helix loop helix protein HEIR 1|Helix loop helix protein HEIR1|Helix-loop-helix protein HEIR-1|HLH642|ID 3|ID like protein inhibitor HLH 1R21|ID like protein inhibitor HLH 462|ID-like protein inhibitor HLH 1R21|ID3|ID3_HUMAN|Inhibitor of DNA binding 3|Inhibitor of DNA binding 3 dominant negative helix loop helix protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q02535
- Q02535
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together