Anti-MAD2L2 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MAD2L2
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-804
- 77842-803
- Antigen symbol:
- MAD2L2
- MAD2L2
- Antigen name:
- Mitotic spindle assembly checkpoint protein MAD2B protein (1-211AA)
- Mitotic spindle assembly checkpoint protein MAD2B protein (1-211AA)
- Antigen synonyms:
- Homolog of REV7 S cerevisiae|hREV7|MAD2 (mitotic arrest deficient yeast, homolog) like 2|MAD2 homolog|MAD2 like 2|MAD2 mitotic arrest deficient like 2|MAD2-like protein 2|MAD2B|Mad2l2|MD2L2_HUMAN|Mitotic Arrest Deficient 2 L2|Mitotic arrest deficient 2-like protein 2|Mitotic arrest deficient homolog like 2|Mitotic arrest deficient like 2 (yeast)|Mitotic arrest deficient yeast homolog|Mitotic spindle assembly checkpoint protein MAD2B|Polymerase (DNA directed) zeta 2 accessory subunit|POLZ2|REV 7|REV7|REV7 homolog|Weakly similar to Mitotic MAD2 protein (S cerevisiae)
- Homolog of REV7 S cerevisiae|hREV7|MAD2 (mitotic arrest deficient yeast, homolog) like 2|MAD2 homolog|MAD2 like 2|MAD2 mitotic arrest deficient like 2|MAD2-like protein 2|MAD2B|Mad2l2|MD2L2_HUMAN|Mitotic Arrest Deficient 2 L2|Mitotic arrest deficient 2-like protein 2|Mitotic arrest deficient homolog like 2|Mitotic arrest deficient like 2 (yeast)|Mitotic arrest deficient yeast homolog|Mitotic spindle assembly checkpoint protein MAD2B|Polymerase (DNA directed) zeta 2 accessory subunit|POLZ2|REV 7|REV7|REV7 homolog|Weakly similar to Mitotic MAD2 protein (S cerevisiae)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UI95
- Q9UI95
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together