Anti-entH Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: entH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Staphylococcus aureus
Specifications
- Catalog No:
- 77817-170
- 77817-169
- Antigen symbol:
- entH
- entH
- Antigen name:
- Recombinant Staphylococcus aureus Enterotoxin type H protein (25-241AA)
- Recombinant Staphylococcus aureus Enterotoxin type H protein (25-241AA)
- Antigen synonyms:
- entH|sehEnterotoxin type H|SEH
- entH|sehEnterotoxin type H|SEH
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Staphylococcus aureus
- Staphylococcus aureus
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0A0M0
- P0A0M0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together