Anti-PIK3R4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PIK3R4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-565
- 77834-566
- Antigen symbol:
- PIK3R4
- PIK3R4
- Antigen name:
- Phosphoinositide 3-kinase regulatory subunit 4 protein (1300-1358AA)
- Phosphoinositide 3-kinase regulatory subunit 4 protein (1300-1358AA)
- Antigen synonyms:
- MGC102700|P150|Phosphatidylinositol 3 kinase associated p150|Phosphatidylinositol 3 kinase regulatory subunit polypeptide 4|Phosphatidylinositol 3 kinase regulatory subunit polypeptide 4 p150|Phosphoinositide 3 kinase adaptor protein|Phosphoinositide 3 kinase regulatory subunit 4|Phosphoinositide 3 kinase regulatory subunit 4 p150|Phosphoinositide 3 kinase regulatory subunit polypeptide 4|Phosphoinositide 3 kinase regulatory subunit polypeptide 4 p150|Phosphoinositide 3-kinase adaptor protein|Phosphoinositide 3-kinase regulatory subunit 4|PI3 kinase p150 subunit|PI3-kinase p150 subunit|PI3-kinase regulatory subunit 4|PI3R4_HUMAN|PIK3R 4|PIK3R4|VPS 15|VPS15
- MGC102700|P150|Phosphatidylinositol 3 kinase associated p150|Phosphatidylinositol 3 kinase regulatory subunit polypeptide 4|Phosphatidylinositol 3 kinase regulatory subunit polypeptide 4 p150|Phosphoinositide 3 kinase adaptor protein|Phosphoinositide 3 kinase regulatory subunit 4|Phosphoinositide 3 kinase regulatory subunit 4 p150|Phosphoinositide 3 kinase regulatory subunit polypeptide 4|Phosphoinositide 3 kinase regulatory subunit polypeptide 4 p150|Phosphoinositide 3-kinase adaptor protein|Phosphoinositide 3-kinase regulatory subunit 4|PI3 kinase p150 subunit|PI3-kinase p150 subunit|PI3-kinase regulatory subunit 4|PI3R4_HUMAN|PIK3R 4|PIK3R4|VPS 15|VPS15
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99570
- Q99570
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together