Anti-AURKC Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:15 to 1:50
Type:
Antigen: AURKC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Specifications
- Catalog No:
- 77819-988
- 77819-989
- Antigen symbol:
- AURKC
- AURKC
- Antigen name:
- Synthetic peptide of AURKC
- Synthetic peptide of AURKC
- Antigen synonyms:
- AURKC|AIE 2|Aie1|AIE2|AIK 3|AIK3|ARK 3|ARK-3|ARK3|Aur C|AurC|Aurkc|AURKC_HUMAN|aurora 3|Aurora C|Aurora kinase C|aurora related kinase 3|Aurora-related kinase 3|Aurora/Ipl1 related kinase 3|Aurora/Ipl1-related kinase 3|Aurora/Ipl1/Eg2 protein 2|EC 2.7.11.1|Serine threonine protein kinase 13|serine/threonine kinase 13 (aurora/IPL1 like)|serine/threonine protein kinase aurora C|Serine/threonine-protein kinase 13|Serine/threonine-protein kinase aurora-C|SPGF5|STK13
- AURKC|AIE 2|Aie1|AIE2|AIK 3|AIK3|ARK 3|ARK-3|ARK3|Aur C|AurC|Aurkc|AURKC_HUMAN|aurora 3|Aurora C|Aurora kinase C|aurora related kinase 3|Aurora-related kinase 3|Aurora/Ipl1 related kinase 3|Aurora/Ipl1-related kinase 3|Aurora/Ipl1/Eg2 protein 2|EC 2.7.11.1|Serine threonine protein kinase 13|serine/threonine kinase 13 (aurora/IPL1 like)|serine/threonine protein kinase aurora C|Serine/threonine-protein kinase 13|Serine/threonine-protein kinase aurora-C|SPGF5|STK13
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UQB9
- Q9UQB9
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together