Anti-RNF133 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: RNF133
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-654
- 77833-653
- Antigen symbol:
- RNF133
- RNF133
- Antigen name:
- E3 ubiquitin-protein ligase RNF133 protein (65-167AA)
- E3 ubiquitin-protein ligase RNF133 protein (65-167AA)
- Antigen synonyms:
- E3 ubiquitin protein ligase RNF133|E3 ubiquitin-protein ligase RNF133|Goliath-related E3 ubiquitin-protein ligase 2|Greul2|MGC27072|RING finger protein 133|RN133_HUMAN|RNF 133|Rnf133
- E3 ubiquitin protein ligase RNF133|E3 ubiquitin-protein ligase RNF133|Goliath-related E3 ubiquitin-protein ligase 2|Greul2|MGC27072|RING finger protein 133|RN133_HUMAN|RNF 133|Rnf133
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WVZ7
- Q8WVZ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together