Anti-SCN2A Rabbit Polyclonal Antibody
Catalog # 77807-451
Supplier: AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Synthesized peptide derived from the Internal region of Na+ CP type IIa.
- Antigen symbol:SCN2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:50 µg
- Form:Liquid
- Gene ID:Q99250
- Antigen synonyms:HBSC II|NAC2|Scn2a|SCN2A_HUMAN|SCN2A1|SCN2A2|Sodium channel protein brain II subunit alpha|Sodium channel protein type 2 subunit alpha|Sodium channel protein type II subunit alpha|Sodium channel protein, brain II subunit alpha|Voltage gated sodium channel subunit alpha Nav1.2|Voltage-gated sodium channel subunit alpha Nav1.2
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: SCN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat