Anti-IKZF4 Rabbit Polyclonal Antibody
Catalog # 77808-055
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from the C-terminal region of Eos.
- Antigen symbol:IKZF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9H2S9
- Antigen synonyms:EOS| Eos protein| IKAROS family zinc finger 4 (Eos)| IKAROS family zinc finger 4| Ikaros family zinc finger protein 4| IKZF4| IKZF4_HUMAN| KIAA1782| Zinc finger protein Eos (IKAROS family zinc finger protein 4)| Zinc finger protein Eos| Zinc finger protein| subfamily 1A| 4 (Eos)| member 4| Zinc finger transcription factor Eos| ZNFN1A4
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:40000
Type:
Antigen: IKZF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat