Anti-SLC28A1 Rabbit Recombinant Polyclonal Antibody (Biotin)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC28A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-970
- 77799-971
- Antigen symbol:
- SLC28A1
- SLC28A1
- Antigen name:
- Sodium/nucleoside cotransporter 1 protein (1-80AA)
- Sodium/nucleoside cotransporter 1 protein (1-80AA)
- Antigen synonyms:
- CNT 1|CNT1|Concentrative nucleoside transporter 1|hCNT1|Na(+)/nucleoside cotransporter 1|S28A1_HUMAN|SLC28A1|Sodium-coupled nucleoside transporter 1|Sodium/nucleoside cotransporter 1|solute carrier family 28 concentrative nucleoside transporter member 1|Solute carrier family 28 member 1
- CNT 1|CNT1|Concentrative nucleoside transporter 1|hCNT1|Na(+)/nucleoside cotransporter 1|S28A1_HUMAN|SLC28A1|Sodium-coupled nucleoside transporter 1|Sodium/nucleoside cotransporter 1|solute carrier family 28 concentrative nucleoside transporter member 1|Solute carrier family 28 member 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00337
- O00337
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together