Anti-TBPL1 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TBPL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-984
- 77801-983
- Antigen symbol:
- TBPL1
- TBPL1
- Antigen name:
- TATA box-binding protein-like protein 1 protein (1-185AA)
- TATA box-binding protein-like protein 1 protein (1-185AA)
- Antigen synonyms:
- 21 kDa TBP-like protein|21kDa TBP like protein|Second TBP of unique DNA|Second TBP of unique DNA protein|STUD|TATA box binding protein like protein 1|TATA box binding protein-related factor 2|TATA box-binding protein-like protein 1|TATA box-binding protein-related factor 2|TBP like 1|TBP like protein|TBP related factor 2|TBP-like factor|TBP-like protein 1|TBP-related factor 2|TBP-related protein|tbpl1|TBPL1_HUMAN|TLF|TLP|TRF2
- 21 kDa TBP-like protein|21kDa TBP like protein|Second TBP of unique DNA|Second TBP of unique DNA protein|STUD|TATA box binding protein like protein 1|TATA box binding protein-related factor 2|TATA box-binding protein-like protein 1|TATA box-binding protein-related factor 2|TBP like 1|TBP like protein|TBP related factor 2|TBP-like factor|TBP-like protein 1|TBP-related factor 2|TBP-related protein|tbpl1|TBPL1_HUMAN|TLF|TLP|TRF2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P62380
- P62380
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together