Order Entry
Canada
ContactUsLinkComponent
 
Anti-BRINP3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
 
undefined
Anti-BRINP3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product

 

  • Catalog No:
  • 77830-767
  • 77830-766
  • Antigen symbol:
  • BRINP3
  • BRINP3
  • Antigen name:
  • BMP/retinoic acid-inducible neural-specific protein 3 protein (389-487AA)
  • BMP/retinoic acid-inducible neural-specific protein 3 protein (389-487AA)
  • Antigen synonyms:
  • BRINP3|DBCCR1L|DBCCR1L1|FAM5CBMP/retinoic acid-inducible neural-specific protein 3|DBCCR1-like protein 1
  • BRINP3|DBCCR1L|DBCCR1L1|FAM5CBMP/retinoic acid-inducible neural-specific protein 3|DBCCR1-like protein 1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q76B58
  • Q76B58
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together