Anti-ZSCAN16 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: ZSCAN16
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77835-574
- 77835-573
- Antigen symbol:
- ZSCAN16
- ZSCAN16
- Antigen name:
- Zinc finger and SCAN domain-containing protein 16 protein (1-348AA)
- Zinc finger and SCAN domain-containing protein 16 protein (1-348AA)
- Antigen synonyms:
- ZSCAN16|ZNF392|ZNF435|Zinc finger and SCAN domain-containing protein 16|Zinc finger protein 392|Zinc finger protein 435
- ZSCAN16|ZNF392|ZNF435|Zinc finger and SCAN domain-containing protein 16|Zinc finger protein 392|Zinc finger protein 435
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H4T2
- Q9H4T2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together