Anti-BTG4 Rabbit Recombinant Polyclonal Antibody
77839-954
New Product
- Antigen name:Protein BTG4 protein (1-206AA)
- Antigen symbol:BTG4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NY30
- Antigen synonyms:B cell translocation gene 4| BTG family member 4| Btg4| BTG4_HUMAN| MGC33003| PC3B | Protein BTG4| Protein PC3b
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: BTG4
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse