Order Entry
Canada
ContactUsLinkComponent
Anti-ACS8 Rabbit Recombinant Polyclonal Antibody (FITC)
Anti-ACS8 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-ACS8 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier:  AFG Bioscience
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 77837-838
  • 77837-837
  • Antigen symbol:
  • ACS8
  • ACS8
  • Antigen name:
  • Recombinant Arabidopsis thaliana 1-aminocyclopropane-1-carboxylate synthase 8 protein (1-469AA)
  • Recombinant Arabidopsis thaliana 1-aminocyclopropane-1-carboxylate synthase 8 protein (1-469AA)
  • Antigen synonyms:
  • ACS8|At4g37770|T28I19.501-aminocyclopropane-1-carboxylate synthase 8|ACC synthase 8|EC 4.4.1.14|S-adenosyl-L-methionine methylthioadenosine-lyase 8
  • ACS8|At4g37770|T28I19.501-aminocyclopropane-1-carboxylate synthase 8|ACC synthase 8|EC 4.4.1.14|S-adenosyl-L-methionine methylthioadenosine-lyase 8
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • A. thaliana
  • A. thaliana
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9T065
  • Q9T065
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, protein G purified
  • >95%, protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together