Anti-HSD17B10 Rabbit Recombinant Polyclonal Antibody
77835-114
New Product
- Antigen name:3-hydroxyacyl-CoA dehydrogenase type-2 protein (2-261AA)
- Antigen symbol:HSD17B10
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q99714
- Antigen synonyms:17 beta hydroxysteroid dehydrogenase 10|17 beta hydroxysteroid dehydrogenase type 10|17-beta-HSD 10|17-beta-hydroxysteroid dehydrogenase 10|17b HSD10|3 hydroxy 2 methylbutyryl CoA dehydrogenase|3 hydroxyacyl CoA dehydrogenase type 2|3 hydroxyacyl CoA dehydrogenase type II|3-hydroxy-2-methylbutyryl-CoA dehydrogenase|3-hydroxyacyl-CoA dehydrogenase type II|3-hydroxyacyl-CoA dehydrogenase type-2|AB binding alcohol dehydrogenase|ABAD|Ads9|Amyloid beta binding polypeptide|Amyloid beta peptide binding alcohol dehydrogenase|Amyloid beta peptide binding protein|CAMR|DUPXp11.22|Endoplasmic Reticulum Amyloid Binding Protein|Endoplasmic reticulum associated amyloid beta peptide binding protein|Endoplasmic reticulum-associated amyloid beta-peptide-binding protein|ER associated amyloid beta-binding protein|ERAB|HADH 2|HADH2|HCD 2|HCD2|HCD2_HUMAN|Hsd17b10|Hydroxyacyl CoA Dehydrogenase type II|Hydroxyacyl Coenzyme A dehydrogenase type II|Hydroxysteroid (17 beta) dehydrogenase 10|Mental retardation X linked syndromic 11|MHBD|Mitochondrial L3 Hydroxyacyl CoA Dehydrogenase|Mitochondrial ribonuclease P protein 2|Mitochondrial RNase P protein 2|MRPP2|MRX17|SCHAD|SDR5C1|Short chain dehydrogenase/reductase family 5C member 1|Short chain L 3 hydroxyacyl CoA dehydrogenase type 2|Short chain type dehydrogenase/reductase XH98G2|Short-chain type dehydrogenase/reductase XH98G2|Type 10 17b HSD|Type 10 17beta hydroxysteroid dehydrogenase|Type II HADH|XH98G2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IF: 1:50 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: HSD17B10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human