Anti-CRHR2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CRHR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-192
- 77825-191
- Antigen symbol:
- CRHR2
- CRHR2
- Antigen name:
- Corticotropin-releasing factor receptor 2 protein (20-108AA)
- Corticotropin-releasing factor receptor 2 protein (20-108AA)
- Antigen synonyms:
- Corticotropin releasing hormone receptor 2|Corticotropin-releasing factor receptor 2|Corticotropin-releasing hormone receptor 2|CRF 2|CRF 2R|CRF R2|CRF RB|CRF-R-2|CRF-R2|CRF2|CRF2 receptor beta isoform|CRF2R|CRFR 2|CRFR-2|CRFR2|CRFR2_HUMAN|CRH 2R|CRH R2|CRH-R-2|CRH-R2|CRH2R|CRHR 2|Crhr2|HM CRF
- Corticotropin releasing hormone receptor 2|Corticotropin-releasing factor receptor 2|Corticotropin-releasing hormone receptor 2|CRF 2|CRF 2R|CRF R2|CRF RB|CRF-R-2|CRF-R2|CRF2|CRF2 receptor beta isoform|CRF2R|CRFR 2|CRFR-2|CRFR2|CRFR2_HUMAN|CRH 2R|CRH R2|CRH-R-2|CRH-R2|CRH2R|CRHR 2|Crhr2|HM CRF
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13324
- Q13324
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together