Anti-ATRIP Rabbit Polyclonal Antibody
Catalog # 77809-412
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Synthesized peptide derived from ATRIP around the phosphorylation site of S224.
- Antigen symbol:ATRIP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q8WXE1
- Antigen synonyms:AGS1|ATIP|ATM and Rad3 related interacting protein|ATM and Rad3-related-interacting protein|ATR interacting protein|ATR-interacting protein|Atrip|ATRIP_HUMAN|DKFZp762J2115|FLJ12343|MGC20625|MGC21482|MGC26740
- Storage buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, ELISA: 1:10000
Type:
Antigen: ATRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human