Anti-SS18L1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SS18L1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-083
- 77801-082
- Antigen symbol:
- SS18L1
- SS18L1
- Antigen name:
- Calcium-responsive transactivator protein (47-396AA)
- Calcium-responsive transactivator protein (47-396AA)
- Antigen synonyms:
- Calcium-responsive transactivator|CREST|CREST_HUMAN|KIAA0693|LP2261|MGC26711|MGC78386|sarcoma translocation gene on chromosome18like1|SS18|SS18-like protein 1|SS18L1|SS18L1 nBAF chromatin remodeling complex|SSXT|synovial sarcoma translocation|synovial sarcoma translocation gene on chromosome 18-like 1|SYT homolog 1
- Calcium-responsive transactivator|CREST|CREST_HUMAN|KIAA0693|LP2261|MGC26711|MGC78386|sarcoma translocation gene on chromosome18like1|SS18|SS18-like protein 1|SS18L1|SS18L1 nBAF chromatin remodeling complex|SSXT|synovial sarcoma translocation|synovial sarcoma translocation gene on chromosome 18-like 1|SYT homolog 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75177
- O75177
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together