Anti-VACWR062 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VACWR062
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Vaccinia virus
Specifications
- Catalog No:
- 77815-563
- 77815-562
- Antigen symbol:
- VACWR062
- VACWR062
- Antigen name:
- Recombinant Vaccinia virus Protein E6 protein (180-429AA)
- Recombinant Vaccinia virus Protein E6 protein (180-429AA)
- Antigen synonyms:
- VACWR062|E6RProtein E6
- VACWR062|E6RProtein E6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Vaccinia virus (VACV)
- Vaccinia virus (VACV)
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P21607
- P21607
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together