Anti-NT5C3A Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NT5C3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-079
- 77837-078
- Antigen symbol:
- NT5C3A
- NT5C3A
- Antigen name:
- Cytosolic 5\\\'-nucleotidase 3A protein (37-336AA)
- Cytosolic 5\\\'-nucleotidase 3A protein (37-336AA)
- Antigen synonyms:
- 5NT3A_HUMAN|cN III |cN-III|cNIII |Cytosolic 5' nucleotidase 3|Cytosolic 5' nucleotidase III|Cytosolic 5''-nucleotidase 3|Cytosolic 5''-nucleotidase 3A|Cytosolic 5''-nucleotidase III|NT5C3|Nt5c3a|p36|P5''N-1|P5'N 1 |P5'N1 |P5N 1 |P5N-1|P5N1 |PN I |PN-I|PNI |PSN1 |Pyrimidine 5' nucleotidase 1|Pyrimidine 5''-nucleotidase 1|UMPH |UMPH1 |Uridine 5' monophosphate hydrolase 1|Uridine 5''-monophosphate hydrolase 1
- 5NT3A_HUMAN|cN III |cN-III|cNIII |Cytosolic 5' nucleotidase 3|Cytosolic 5' nucleotidase III|Cytosolic 5''-nucleotidase 3|Cytosolic 5''-nucleotidase 3A|Cytosolic 5''-nucleotidase III|NT5C3|Nt5c3a|p36|P5''N-1|P5'N 1 |P5'N1 |P5N 1 |P5N-1|P5N1 |PN I |PN-I|PNI |PSN1 |Pyrimidine 5' nucleotidase 1|Pyrimidine 5''-nucleotidase 1|UMPH |UMPH1 |Uridine 5' monophosphate hydrolase 1|Uridine 5''-monophosphate hydrolase 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H0P0
- Q9H0P0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together