Anti-TMEM59 Rabbit Recombinant Polyclonal Antibody
Catalog # 77811-086
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:Transmembrane protein 59 protein (59-201AA)
- Antigen symbol:TMEM59
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9BXS4
- Antigen synonyms:C1orf8|HSPC001|Liver membrane-bound protein|TMEM59|TMEM59 transmembrane protein 59|TMM59_HUMAN|Transmembrane protein 59
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: TMEM59
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human