Anti-FOXE3 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Type:
Antigen: FOXE3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77823-427
- 77823-426
- Antigen symbol:
- FOXE3
- FOXE3
- Antigen name:
- Forkhead box protein E3 protein (4-74AA)
- Forkhead box protein E3 protein (4-74AA)
- Antigen synonyms:
- Drosphilia Forkhead Homolog Like 12|FKHL 12|FKHL12|Forkhead Box E3|Forkhead box protein E3|Forkhead Related Activator 8|Forkhead related protein FKHL12|Forkhead-related protein FKHL12|Forkhead-related transcription factor 8|Foxe3|FOXE3 forkhead box E3|FOXE3_HUMAN|FREAC 8|FREAC-8|FREAC8
- Drosphilia Forkhead Homolog Like 12|FKHL 12|FKHL12|Forkhead Box E3|Forkhead box protein E3|Forkhead Related Activator 8|Forkhead related protein FKHL12|Forkhead-related protein FKHL12|Forkhead-related transcription factor 8|Foxe3|FOXE3 forkhead box E3|FOXE3_HUMAN|FREAC 8|FREAC-8|FREAC8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13461
- Q13461
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together