Anti-S1PR2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: S1PR2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77798-597
- 77798-598
- Antigen symbol:
- S1PR2
- S1PR2
- Antigen name:
- Sphingosine 1-phosphate receptor 2 protein (293-353AA)
- Sphingosine 1-phosphate receptor 2 protein (293-353AA)
- Antigen synonyms:
- AGR16|EDG 5|EDG5|Endothelial Cell Differentiation Gene 5|Endothelial differentiation G protein coupled receptor 5|Endothelial differentiation G-protein coupled receptor 5|Endothelial differentiation sphingolipid G protein coupled receptor 5|Gpcr13|H218|LPb2|Lysophospholipid receptor B2|S1P receptor 2|S1P receptor Edg 5|S1P receptor Edg-5|S1P receptor EDG5|S1P2|S1pr2|S1PR2_HUMAN|Sphingosine 1 phosphate receptor Edg 5|Sphingosine 1-phosphate receptor 2|Sphingosine 1-phosphate receptor Edg-5
- AGR16|EDG 5|EDG5|Endothelial Cell Differentiation Gene 5|Endothelial differentiation G protein coupled receptor 5|Endothelial differentiation G-protein coupled receptor 5|Endothelial differentiation sphingolipid G protein coupled receptor 5|Gpcr13|H218|LPb2|Lysophospholipid receptor B2|S1P receptor 2|S1P receptor Edg 5|S1P receptor Edg-5|S1P receptor EDG5|S1P2|S1pr2|S1PR2_HUMAN|Sphingosine 1 phosphate receptor Edg 5|Sphingosine 1-phosphate receptor 2|Sphingosine 1-phosphate receptor Edg-5
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95136
- O95136
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together