Anti-GTF2H2C Rabbit Recombinant Polyclonal Antibody
Catalog # 77830-855
Supplier: AFG Bioscience
New Product
Specifications
- Antigen name:General transcription factor IIH subunit 2-like protein (1-260AA)
- Antigen symbol:GTF2H2C
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q6P1K8
- Antigen synonyms:GTF2H2C|GTF2H2C_2|GTF2H2D|General transcription factor IIH subunit 2-like protein|General transcription factor IIH polypeptide 2-like protein
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: GTF2H2C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human