Anti-TERF2 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: TERF2
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77824-118
- 77824-117
- Antigen symbol:
- TERF2
- TERF2
- Antigen name:
- Telomeric repeat-binding factor 2 protein (78-238AA)
- Telomeric repeat-binding factor 2 protein (78-238AA)
- Antigen synonyms:
- Telomeric DNA binding protein|Telomeric DNA-binding protein|Telomeric repeat binding factor 2|Telomeric repeat binding protein 2|Telomeric repeat-binding factor 2|TERF 2|Terf2|TERF2_HUMAN|TRBF 2|TRBF2|TRF 2|TRF2|TTAGGG repeat binding factor 2|TTAGGG repeat-binding factor 2
- Telomeric DNA binding protein|Telomeric DNA-binding protein|Telomeric repeat binding factor 2|Telomeric repeat binding protein 2|Telomeric repeat-binding factor 2|TERF 2|Terf2|TERF2_HUMAN|TRBF 2|TRBF2|TRF 2|TRF2|TTAGGG repeat binding factor 2|TTAGGG repeat-binding factor 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q15554
- Q15554
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together