Anti-Klk1b22 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: Klk1b22
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77815-578
- 77815-579
- Antigen symbol:
- Klk1b22
- Klk1b22
- Antigen name:
- Recombinant Mouse Kallikrein 1-related peptidase b22 protein (25-259AA)
- Recombinant Mouse Kallikrein 1-related peptidase b22 protein (25-259AA)
- Antigen synonyms:
- Klk1b22|Klk-22|Klk22|Kallikrein 1-related peptidase b22|EC 3.4.21.35|Beta-NGF-endopeptidase|Epidermal growth factor-binding protein type A|EGF-BP A|Glandular kallikrein K22|mGK-22|Nerve growth factor beta chain endopeptidase|Tissue kallikrein 22
- Klk1b22|Klk-22|Klk22|Kallikrein 1-related peptidase b22|EC 3.4.21.35|Beta-NGF-endopeptidase|Epidermal growth factor-binding protein type A|EGF-BP A|Glandular kallikrein K22|mGK-22|Nerve growth factor beta chain endopeptidase|Tissue kallikrein 22
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15948
- P15948
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together