Anti-TRIM24 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM24
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-535
- 77803-535
- Antigen symbol:
- TRIM24
- TRIM24
- Antigen name:
- Transcription intermediary factor 1-alpha protein (891-1012AA)
- Transcription intermediary factor 1-alpha protein (891-1012AA)
- Antigen synonyms:
- E3 ubiquitin protein ligase TRIM24|E3 ubiquitin-protein ligase Trim24|hTIF1|PTC6|RING finger protein 82|RNF82|TF1A|TIF1 alpha|TIF1|TIF1-alpha|TIF1A|TIF1A_HUMAN|TIF1ALPHA|Transcription intermediary factor 1-alpha|Transcriptional intermediary factor 1 alpha|Transcriptional intermediary factor 1|Trim24|Tripartite motif containing 24|Tripartite motif-containing protein 24
- E3 ubiquitin protein ligase TRIM24|E3 ubiquitin-protein ligase Trim24|hTIF1|PTC6|RING finger protein 82|RNF82|TF1A|TIF1 alpha|TIF1|TIF1-alpha|TIF1A|TIF1A_HUMAN|TIF1ALPHA|Transcription intermediary factor 1-alpha|Transcriptional intermediary factor 1 alpha|Transcriptional intermediary factor 1|Trim24|Tripartite motif containing 24|Tripartite motif-containing protein 24
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O15164
- O15164
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together