Order Entry
Canada
ContactUsLinkComponent
Anti-PITX2 Rabbit Recombinant Polyclonal Antibody (HRP)
Anti-PITX2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-PITX2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77835-047
  • 77835-048
  • Antigen symbol:
  • PITX2
  • PITX2
  • Antigen name:
  • Pituitary homeobox 2 protein (40-154AA)
  • Pituitary homeobox 2 protein (40-154AA)
  • Antigen synonyms:
  • All1 responsive gene 1|ALL1 responsive protein ARP1|ALL1-responsive protein ARP1|ARP 1|ARP1|Brx 1|Brx1|Homeobox protein PITX2|IDG 2|IDG2|IGDS 2|IGDS|IGDS2|IHG 2|IHG2|IRID 2|IRID2|MGC111022|MGC20144|Otlx 2|Otlx2|Paired like homeodomain transcription factor 2|Paired-like homeodomain transcription factor 2|Pituitary homeo box 2|Pituitary homeobox 2|PITX 2|pitx2|PITX2_HUMAN|PTX 2|PTX2|RGS|RIEG 1|RIEG|Rieg bicoid related homeobox transcription factor 1|RIEG bicoid related homeobox transcription factor|RIEG bicoid-related homeobox transcription factor|RIEG1|RS|Solurshin
  • All1 responsive gene 1|ALL1 responsive protein ARP1|ALL1-responsive protein ARP1|ARP 1|ARP1|Brx 1|Brx1|Homeobox protein PITX2|IDG 2|IDG2|IGDS 2|IGDS|IGDS2|IHG 2|IHG2|IRID 2|IRID2|MGC111022|MGC20144|Otlx 2|Otlx2|Paired like homeodomain transcription factor 2|Paired-like homeodomain transcription factor 2|Pituitary homeo box 2|Pituitary homeobox 2|PITX 2|pitx2|PITX2_HUMAN|PTX 2|PTX2|RGS|RIEG 1|RIEG|Rieg bicoid related homeobox transcription factor 1|RIEG bicoid related homeobox transcription factor|RIEG bicoid-related homeobox transcription factor|RIEG1|RS|Solurshin
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • HRP (Horseradish Peroxidase)
  • HRP (Horseradish Peroxidase)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q99697
  • Q99697
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage temperature:
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together