Anti-POU3F1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: POU3F1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77794-887
- 77794-888
- Antigen symbol:
- POU3F1
- POU3F1
- Antigen name:
- POU domain, class 3, transcription factor 1 protein (59-257AA)
- POU domain, class 3, transcription factor 1 protein (59-257AA)
- Antigen synonyms:
- class 3|Oct-6|OCT6|Octamer-binding protein 6|Octamer-binding transcription factor 6|OTF-6|OTF6|PO3F1_HUMAN|POU class 3 homeobox 1|POU domain|POU domain class 3 transcription factor 1|POU domain transcription factor SCIP|pou3f1|SCIP|transcription factor 1
- class 3|Oct-6|OCT6|Octamer-binding protein 6|Octamer-binding transcription factor 6|OTF-6|OTF6|PO3F1_HUMAN|POU class 3 homeobox 1|POU domain|POU domain class 3 transcription factor 1|POU domain transcription factor SCIP|pou3f1|SCIP|transcription factor 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q03052
- Q03052
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together