Anti-METTL2A Rabbit Recombinant Polyclonal Antibody
77834-134
New Product
- Antigen name:Methyltransferase-like protein 2A protein (1-242AA)
- Antigen symbol:METTL2A
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96IZ6
- Antigen synonyms:METTL2A| METTL2| HSPC266| tRNA N(3)-methylcytidine methyltransferase METTL2A| EC 2.1.1.-| Methyltransferase-like protein 2A
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on Blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:2000 to 1:5000
Type:
Antigen: METTL2A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse