Anti-DDX18 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:200 to 1:500
Type:
Antigen: DDX18
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-001
- 77841-002
- Antigen symbol:
- DDX18
- DDX18
- Antigen name:
- ATP-dependent RNA helicase DDX18 protein (32-110AA)
- ATP-dependent RNA helicase DDX18 protein (32-110AA)
- Antigen synonyms:
- ATP dependent RNA helicase DDX18|ATP-dependent RNA helicase DDX18|DDX 18|DDX18|DDX18_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 18|DEAD box polypeptide 18|DEAD box protein 18|FLJ33908|MrDb|Myc regulated DEAD box protein|Myc-regulated DEAD box protein
- ATP dependent RNA helicase DDX18|ATP-dependent RNA helicase DDX18|DDX 18|DDX18|DDX18_HUMAN|DEAD (Asp Glu Ala Asp) box polypeptide 18|DEAD box polypeptide 18|DEAD box protein 18|FLJ33908|MrDb|Myc regulated DEAD box protein|Myc-regulated DEAD box protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVP1
- Q9NVP1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together