Anti-fim3 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: fim3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bordetella pertussis
Specifications
- Catalog No:
- 77815-421
- 77815-422
- Antigen symbol:
- fim3
- fim3
- Antigen name:
- Recombinant Bordetella pertussis Serotype 3 fimbrial subunit protein (26-204AA)
- Recombinant Bordetella pertussis Serotype 3 fimbrial subunit protein (26-204AA)
- Antigen synonyms:
- fim3|BP1568|Serotype 3 fimbrial subunit
- fim3|BP1568|Serotype 3 fimbrial subunit
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- B. pertussis
- B. pertussis
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P17835
- P17835
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together